missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltriethoxysilane, 98%

Product Code. 11323866
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323866 100 g 100g
11313866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323866 Supplier Thermo Scientific Alfa Aesar Supplier No. L16965.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2943-73-9 | C16H36O3Si | 304.546 g/mol

n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2943-73-9
Molecular Formula C16H36O3Si
Molecular Weight (g/mol) 304.546
MDL Number MFCD00069132
InChI Key BAAAEEDPKUHLID-UHFFFAOYSA-N
Synonym n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy
PubChem CID 76261
IUPAC Name decyl(triethoxy)silane
SMILES CCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 150°C (8mmHg)
Quantity 100 g
Linear Formula CH3(CH2)9Si(OCH2CH3)3
Beilstein 2413155
Flash Point >110°C (230°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 304.55
Percent Purity 98%
Refractive Index 1.422
Sensitivity Moisture sensitive
Density 0.879
Chemical Name or Material n-Decyltriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.