missing translation for 'onlineSavingsMsg'
Learn More

n-Decyldimethylchlorosilane, 97%, Thermo Scientific™

Product Code. 11385587
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385587 50 g 50g
11395587 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385587 Supplier Thermo Scientific Alfa Aesar Supplier No. 042769.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38051-57-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00054168
InChI Key ZLZGHBNDPINFKG-UHFFFAOYSA-N
Synonym chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
PubChem CID 2758020
IUPAC Name chloro-decyl-dimethylsilane
SMILES CCCCCCCCCC[Si](C)(C)Cl

Specifications

Boiling Point 98°C (2mmHg)
Physical Form Liquid
Quantity 50 g
UN Number UN1760
Flash Point 137°C (279°F)
Formula Weight 234.88
Percent Purity 97%
Refractive Index 1.441
Sensitivity Moisture sensitive
Density 0.866 g/mL
Chemical Name or Material n-Decyldimethylchlorosilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.