missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
| CAS | 15163-36-7 |
| Molecular Formula | C15 H33 N O3 S |
| Quantity | 25 g |
| Synonym | 1-Decanaminium, N,N-dimethyl-N-(3-sulfopropyl)-, inner salt, Ammonium, decyldimethyl(3-sulfopropyl)-, hydroxide, inner salt (8CI), 3-(Decyldimethylammonio)propane-1-sulfonate, 3-(Decyldimethylammonio)propanesulfonate, 3-(Decyldimethylammonio)propanesulfonate inner salt, 3-(N-Decyl-N,N-dimethylammonio)-1-propanesulfonate, Anzergent 3-10, DeDAPS, N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, N-Decylsulfobetaine, SB 10, SB 10 (onium compound), SB 3-10, Sulfobetaine 10, Zwittergent 3-10 |
| InChI Formula | InChI=1S/C15H33NO3S/c1-4-5-6-7-8-9-10-11-13-16(2,3)14-12-15-20(17,18)19/h4-15H2,1-3H3 |
| SMILES | CCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| IUPAC Name | 3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate |
| Molecular Weight (g/mol) | 307.49 |
| Formula Weight | 307.2181 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction