missing translation for 'onlineSavingsMsg'
Learn More

n-Decyl-d21-trimethylammonium Bromide, TRC

Product Code. 30381704
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381704 2 mg
30387172 10 mg
30376728 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381704 Supplier Toronto Research Chemicals Supplier No. TRCD5255392MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1515861-67-2
Molecular Formula C13H9D21BrN
Molecular Weight (g/mol) 301.42
Synonym 1-Decan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21-aminium, N,N,N-trimethyl-, bromide (1:1) (ACI), Decyl-d21-trimethylammonium bromide, Decyltrimethylammonium-D21 (decyl-D21) bromide, Decyltrimethylammonium-d21 bromide, N,N,N-Trimethyl-1-decan-d21-a
IUPAC Name N,N,N-trimethyldecan-1-aminium-d21 bromide
SMILES C(C(C(C(C(C(C(C(C([2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])(C([N+](C)(C)C)([2H])[2H])([2H])[2H].[Br-]

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C13H30N.BrH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1H/q+1;/p-1/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2;
Formula Weight 300.29
Chemical Name or Material Decyltrimethylammonium-D21 (decyl-D21) Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.