missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Decyl-d21-trimethylammonium Bromide, CDN
Specifications
Specifications
| Chemical Name or Material | Decyltrimethylammonium-D21 (decyl-D21) Bromide |
| CAS | 1515861-67-2 |
| Molecular Formula | C13H9D21BrN |
| Quantity | 0.05 g |
| Synonym | 1-Decan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21-aminium, N,N,N-trimethyl-, bromide (1:1) (ACI), Decyl-d21-trimethylammonium bromide, Decyltrimethylammonium-D21 (decyl-D21) bromide, Decyltrimethylammonium-d21 bromide, N,N,N-Trimethyl-1-decan-d21-a |
| InChI Formula | InChI=1 S/C13H30N.BrH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1 H/q+1;/p-1/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2; |
| SMILES | C(C(C(C(C(C(C(C(C([2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])(C([N+](C)(C)C)([2 H])[2 H])([2 H])[2 H].[Br-] |
| IUPAC Name | N,N,N-trimethyldecan-1-aminium-d21 bromide |
| Molecular Weight (g/mol) | 301.42 |
| Formula Weight | 300.29 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?