missing translation for 'onlineSavingsMsg'
Learn More

N-Decanoyl-N-methylglucamine, TRC

Product Code. 30364179
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364179 250 mg 250mg
30358116 50 mg 50mg
30355597 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364179 Supplier Toronto Research Chemicals Supplier No. TRCD212658250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85261-20-7
Molecular Formula C17H35NO6
Molecular Weight (g/mol) 349.46
Synonym 1-Deoxy-1-[methyl(1-oxodecyl)amino]-D-glucitol, Decanoyl N-methylglucamide, MEGA 10, N-Decanoyl-N-methyl-1-amino-1-deoxy-D-glucitol, N-Methyl-N-decanoylglucamine
IUPAC Name N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
SMILES CCCCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H35NO6/c1-3-4-5-6-7-8-9-10-15(22)18(2)11-13(20)16(23)17(24)14(21)12-19/h13-14,16-17,19-21,23-24H,3-12H2,1-2H3/t13-,14+,16+,17+/m0/s1
Formula Weight 349.2464
Chemical Name or Material N-Decanoyl-N-methylglucamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.