missing translation for 'onlineSavingsMsg'
Learn More

n-Decane, MP Biomedicals™

Product Code. 11430992 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430992 100 mL 100mL
11440992 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430992 Supplier MP Biomedicals Supplier No. 0219512380
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

n-Decane >99%, Clear, Colorless Liquid

  • Decane has been used as a solvent in the preparation of high alkane phase emulsified suspensions (HAPES).
  • It may also be used as a solvent to suspend the CdSe/ZnS nanocrystals during absorbance measurement.
  • Internal standard in the GC analysis of oxalic, malonic, and succinic acids in biological materials.
  • Decane (n-Decane), an aliphatic hydrocarbon, is a reference fuel. Its oxidation over a range of temperature has been studied.
  • Its crystal structure has been investigated at 150K.
  • The cracking of decane can be performed using electrohydrodynamic atomization system (EHDA).

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Melting Point −30°C (Literature)
Color Colorless
Density 0.734g/cm3 at 15°C(Lit.)
Boiling Point 174°C (Literature)
Refractive Index n20/D 1.411(Lit.)
Quantity 100 mL
UN Number UN1760
Formula Weight 142.29
Percent Purity 0.996
Physical Form Liquid
Chemical Name or Material n-Decane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Decane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.