missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Decane, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
Description
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -30°C |
Color | Colorless |
Density | 0.730 g/mL |
Boiling Point | 172°C to 174°C |
Flash Point | 46°C (115°F) |
Assay Percent Range | >99% |
Odor | Petrol-like |
Linear Formula | CH3(CH2)8CH3 |
Refractive Index | 1.4115 |
Quantity | 10 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'