missing translation for 'onlineSavingsMsg'
Learn More

N-Debenzoyl-N-tigloylpaclitaxel (Cephalomannine), Mikromol™

Product Code. 17900466
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17900466 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17900466 Supplier Mikromol Supplier No. MM1489.020025
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Debenzoyl-N-tigloylpaclitaxel (Cephalomannine)
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Cephalomannine
CAS 71610-00-9
Analyte or Component Names Cephalomannine
Shipping Condition Room Temperature
Molecular Formula C45 H53 N O14
Certifications/Compliance ISO 9001
Synonym Paclitaxel Related Compound A, Paclitaxel Imp B (EP), N-Debenzoyl-N-tigloylpaclitaxel, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., Cephalomannine, NSC 318735, Taxol B, Paclitaxel USP Related Compound A, Paclitaxel USP RC A, Benzenepropanoic acid, alpha-hydroxy-beta-[[(2E)-2-methyl-1-oxo-2-buten-1-yl]amino]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (alphaR,betaS)-, Benzenepropanoic acid, alpha-hydroxy-beta-[(2-methyl-1-oxo-2-butenyl)amino]-, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aalpha,4beta,4abeta,6beta,9alpha[alphaR*,betaS*(E)],11alpha,12alpha,12aalpha,12balpha]]-, Benzenepropanoic acid, alpha-hydroxy-beta-[[(2E)-2-methyl-1-oxo-2-butenyl]amino]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (alphaR,betaS)- (9CI), Paclitaxel Related Compound A, Paclitaxel Impurity B
InChI Formula InChI=1S/C45H53NO14/c1-9-23(2)39(52)46-33(27-16-12-10-13-17-27)34(50)41(54)58-29-21-45(55)38(59-40(53)28-18-14-11-15-19-28)36-43(8,30(49)20-31-44(36,22-56-31)60-26(5)48)37(51)35(57-25(4)47)32(24(29)3)42(45,6)7/h9-19,29-31,33-36,38,49-50,55H,20-22H2,1-8H3,(H,46,52)/b23-9+/t29-,30-,31+,33-,34+,35+,36-,38-,43+,44-,45+/m0/s1
SMILES C\C=C(/C)\C(=O)N[C@H]([C@@H](O)C(=O)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@@H]4[C@@]5(CO[C@@H]5C[C@H](O)[C@@]4(C)C(=O)[C@H](OC(=O)C)C(=C1C)C2(C)C)OC(=O)C)c6ccccc6
IUPAC Name (αR,βS)-α-Hydroxy-β-[[(2E)-2-methyl-1-oxo-2-buten-1-yl]amino]benzenepropanoic acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester
Molecular Weight (g/mol) 831.9
Quantity 25 mg
Formula Weight 831.3466 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.