missing translation for 'onlineSavingsMsg'
Learn More

N-deacetylated BMS-202, MedChemExpress

Product Code. 16443686
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443686

Brand: Medchem Express HY19745A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-deacetylated BMS-202 is the deacetylated of BMS-202. BMS-202 is an inhibitor of the PD-1/PD-L1 interaction, mainly used for cancer treatment.

N-deacetylated BMS-202 is the deacetylated of BMS-202. BMS-202 is an inhibitor of the PD-1/PD-L1 interaction, mainly used for cancer treatment.

Specifications

Chemical Name or Material N-deacetylated BMS-202
Physical Form Solid
CAS 2310135-18-1
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C23H27N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 36 mg/mL (95.37 mM)
SMILES NCCNCC1=CC=C(OCC2=C(C)C(C3=CC=CC=C3)=CC=C2)N=C1OC
Molecular Weight (g/mol) 377.48
Formula Weight 377.48
Percent Purity 98.13%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.