missing translation for 'onlineSavingsMsg'
Learn More

N-(Cyanomethyl)pyridinium chloride, 97%

Product Code. 11476877
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476877 10 g 10g
11486877 50 g 50g
11496877 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476877 Supplier Thermo Scientific Alfa Aesar Supplier No. A14327.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17281-59-3 | C7H7ClN2 | 154.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17281-59-3
Molecular Formula C7H7ClN2
Molecular Weight (g/mol) 154.60
MDL Number MFCD00031933
InChI Key HEJOROWXIWLCMS-UHFFFAOYSA-M
Synonym 1-cyanomethyl pyridinium chloride, n-cyanomethyl pyridinium chloride, 1-cyanomethyl pyridin-1-ium chloride, acmc-1chuo, cyanomethylpyridinium chloride, n-cyanomethyl pyridiniumchloride, 2-pyridin-1-ium-1-ylacetonitrile chloride, 2-1-pyridin-1-iumyl acetonitrile chloride, 2-pyridin-1-ium-1-ylethanenitrile chloride
PubChem CID 2734116
IUPAC Name 2-pyridin-1-ium-1-ylacetonitrile;chloride
SMILES [Cl-].N#CC[N+]1=CC=CC=C1

Specifications

Melting Point 173°C to 175°C
Quantity 10 g
UN Number UN3439
Beilstein 3728392
Formula Weight 154.6
Percent Purity 97%
Chemical Name or Material N-(Cyanomethyl)pyridinium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.