missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cyanocyanamide Sodium Salt, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N-Cyanocyanamide Sodium Salt |
| CAS | 1934-75-4 |
| Molecular Formula | C2N3Na |
| Quantity | 10 g |
| Synonym | Dicyanamide, Sodium Salt, Sodium Dicyanamide |
| InChI Formula | InChI=1S/C2N3.Na/c3-1-5-2-4;/q-1;+1 |
| SMILES | [Na+].N#C[N-]C#N |
| IUPAC Name | sodium;dicyanoazanide |
| Molecular Weight (g/mol) | 89.03 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction