missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cbz-L-aspartic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | N-Cbz-L-aspartic Acid |
| CAS | 1152-61-0 |
| Molecular Formula | C12H13NO6 |
| Quantity | 50 g |
| Synonym | N-[(Phenylmethoxy)carbonyl]-L-aspartic Acid, N-Carboxy-L-aspartic Acid N-Benzyl Ester, Benzyloxycarbonyl-L-aspartic Acid, Carbobenzoxy-L-aspartic Acid, Cbz-Asp-OH, N-(Benzyloxycarbonyl)-L-aspartic Acid, N-Benzyloxylcarbonyl-L-aspartic Acid, N-Carbobenzoxy-L-aspartic Acid, N-Carbobenzyloxy-L-aspartic Acid, NSC 88479, NSC 9972 |
| InChI Formula | InChI=1S/C12H13NO6/c14-10(15)6-9(11(16)17)13-12(18)19-7-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,18)(H,14,15)(H,16,17)/t9-/m0/s1 |
| SMILES | OC(=O)C[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)butanedioic acid |
| Molecular Weight (g/mol) | 267.23 |
| Formula Weight | 267.0743 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?