missing translation for 'onlineSavingsMsg'
Learn More

N-Carboxybenzyl-L-serine, TRC

Product Code. 30360704
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360704 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360704 Supplier Toronto Research Chemicals Supplier No. TRCC182415250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Carboxybenzyl-L-serine
CAS 1145-80-8
Molecular Formula C11H13NO5
Quantity 250 mg
Synonym Z-Ser-OH, Z-Serine-OH, (2S)-2-[(Benzyloxycarbonyl)amino]-3-hydroxypropanoic Acid, Benzyloxycarbonyl-L-serine, N-(Benzyloxycarbonyl)-L-serine, N-Carbobenzoxy-L-serine, N-Carbobenzyloxy-L-serine, N-[(Phenylmethoxy)carbonyl]serine, NSC 286604
InChI Formula InChI=1S/C11H13NO5/c13-6-9(10(14)15)12-11(16)17-7-8-4-2-1-3-5-8/h1-5,9,13H,6-7H2,(H,12,16)(H,14,15)/t9-/m0/s1
SMILES OC[C@H](NC(=O)OCc1ccccc1)C(=O)O
IUPAC Name (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
Molecular Weight (g/mol) 239.22
Formula Weight 239.0794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.