missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxyglycine, 98.5%

Product Code. 10328853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10328853 500 g Plastic Bottle 500g
10264180 25 g Plastic Bottle 25g
10476710 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10328853 Supplier Thermo Scientific Acros Supplier No. 108355000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1138-80-3 | C10H11NO4 | 209.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138-80-3
MDL Number MFCD00002691
InChI Key CJUMAFVKTCBCJK-UHFFFAOYSA-N
Synonym n-carbobenzyloxyglycine, n-cbz-glycine, z-gly-oh, carbobenzyloxyglycine, carbobenzoxyglycine, n-benzyloxycarbonylglycine, n-carbobenzoxyglycine, cbz-gly-oh, cbz gly, benzyloxycarbonylglycine
PubChem CID 14349
ChEBI CHEBI:16532
IUPAC Name 2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NCC(=O)O

Specifications

Melting Point 117.0°C to 121.0°C
CAS Min % 98.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Molecular Formula C10H11NO4
Packaging Plastic Bottle
Linear Formula C6H5CH2O2CNHCH2CO2H
Molecular Weight (g/mol) 209.2
Quantity 500 g
Formula Weight 209.2
Percent Purity 98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Carbobenzyloxyglycine, 98.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.