missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-valine, 99+%

Product Code. 10287400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10287400 5 g Glass bottle 5g
10388810 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10287400 Supplier Thermo Scientific Chemicals Supplier No. 275940050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1149-26-4 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-26-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00008922
InChI Key CANZBRDGRHNSGZ-NSHDSACASA-N
Synonym n-carbobenzyloxy-l-valine, n-cbz-l-valine, z-val-oh, cbz-l-valine, n-benzyloxycarbonyl-l-valine, n-carbobenzoxy-l-valine, z-l-val-oh, carbobenzoxy-l-valine, z-valine, carbobenzyloxy-l-valine
PubChem CID 726987
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 58°C to 60°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(NHCO2CH2C6H5)CO2H
Quantity 5 g
Beilstein 06, 1502
Specific Rotation -4
IUPAC Name (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid
Formula Weight 251.28
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-L-valine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.