missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-L-proline, 99%

Product Code. 10080630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10080630 10 g Glass bottle 10g
10667484 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10080630 Supplier Thermo Scientific Acros Supplier No. 108370100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1148-11-4 | C13H15NO4 | 249.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1148-11-4
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.27
MDL Number MFCD00003170
InChI Key JXGVXCZADZNAMJ-NSHDSACASA-N
Synonym z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline
PubChem CID 101987
IUPAC Name (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 76°C to 78°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. D-enantiomer (HPLC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation -60
Solubility Information Solubility in water: negligible. Other solubilities: soluble in most common organic solvents
Formula Weight 249.27
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-Carbobenzyloxy-L-proline, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.