missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-L-methionine, 97%, Thermo Scientific™

Product Code. 11945781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11945781 25 g 25g
11935781 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945781 Supplier Thermo Scientific Acros Supplier No. 442610250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1152-62-1
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.34
MDL Number MFCD00065124
InChI Key FPKHNNQXKZMOJJ-UHFFFAOYNA-N
Synonym z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
PubChem CID 1550857
IUPAC Name (2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 70°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Specific Rotation -27
Formula Weight 283.35
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material N-Carbobenzyloxy-L-methionine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.