missing translation for 'onlineSavingsMsg'
Learn More

N-ε-Carbobenzyloxy-L-lysine, 98%, Thermo Scientific Chemicals

Product Code. 10404355 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10404355 1 g Glass Bottle 1g
10676482 5 g Glass Bottle 5g
10341324 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10404355 Supplier Thermo Scientific Acros Supplier No. 108360010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1155-64-2
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.324
MDL Number MFCD00002638
InChI Key CKGCFBNYQJDIGS-LBPRGKRZSA-N
Synonym h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
PubChem CID 1715626
SMILES C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)O)N

Specifications

Melting Point 259.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
Specific Rotation + 15.50
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Water 0.5% max. (K.F.)
IUPAC Name (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid
Specific Rotation Condition +15.50 (20.00°C c=1,1N HCl)
Quantity 1 g
Formula Weight 280.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.