missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-aspartic acid, 98%

Product Code. 10072200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10072200 25 g 25g
10696134 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072200 Supplier Thermo Scientific Chemicals Supplier No. 172050250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1152-61-0 | C12H13NO6 | 267.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1152-61-0
Molecular Formula C12H13NO6
Molecular Weight (g/mol) 267.23
MDL Number MFCD00002719
InChI Key XYXYXSKSTZAEJW-VIFPVBQESA-N
Synonym z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
PubChem CID 2723942
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 117°C to 119°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH(NHCO2CH2C6H5)CO2H
Quantity 25 g
Specific Rotation + 10.5
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 267.23
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-L-aspartic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.