missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-D-alanine, 98%, Thermo Scientific™

Product Code. 11995561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11995561 1 g 1g
11905571 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995561 Supplier Thermo Scientific Acros Supplier No. 441970010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 26607-51-2
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00063126
InChI Key TYRGLVWXHJRKMT-UHFFFAOYNA-N
Synonym n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine
PubChem CID 70832
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 74°C to 80°C
Color White
Loss on Drying 1.00% max. ()
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation + 15.3
Formula Weight 223.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-D-alanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.