missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxyglycine, TRC

Product Code. 30361789
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361789 25 g -
30357420 100 g 100g
30362126 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361789 Supplier Toronto Research Chemicals Supplier No. TRCC17599525G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138-80-3
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.2
Synonym N-[(Phenylmethoxy)carbonyl]glycine, N-Carboxyglycine N-Benzyl Ester, N-Gly-OH, (Carbobenzoxy)glycine, (Carbobenzyloxy)glycine, 2-(Benzyloxycarbonylamino)acetic Acid, Benzyloxycarbonylglycine, N-(Benzyloxycarbonyl)glycine, N-Cbz-glycine, N-(Carbobenzoxy)glycine, N-(Carbobenzyloxy)glycine, N-(alpha-Carbobenzoxy)glycine, N-Carboxyglycine N-Benzyl Ester, N-[(Phenylmethoxy)carbonyl]glycine, NSC 2526, Nalpha-(Benzyloxycarbonyl)glycine, Z-Glycine, [[(Benzyloxy)carbonyl]amino]acetic Acid
IUPAC Name 2-(phenylmethoxycarbonylamino)acetic acid
SMILES OC(=O)CNC(=O)OCc1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H11NO4/c12-9(13)6-11-10(14)15-7-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,14)(H,12,13)
Formula Weight 209.0688
Chemical Name or Material N-Carbobenzoxyglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.