missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-b-alanine, TRC

Product Code. 30352155
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352155 Supplier Toronto Research Chemicals Supplier No. TRCC17618025G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Carbobenzoxy-beta-alanine
CAS 2304-94-1
Molecular Formula C11H13NO4
Quantity 25 g
Synonym 3-(Benzyloxycarbonylamino)propanoic Acid, 3-(Benzyloxycarbonylamino)propionic Acid, 3-[N-(Benzyloxycarbonyl)amino]propionic Acid, Carbobenzoxy-beta-alanine, N-(Benzyloxycarbonyl)-beta-alanine, N-CBZ-beta-alanine, N-Carbobenzyloxy-beta-alanine, N-[(Phenylmethoxy)carbonyl]-beta-alanine, NSC 17161, NSC 28943, NSC 657842
InChI Formula InChI=1S/C11H13NO4/c13-10(14)6-7-12-11(15)16-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,15)(H,13,14)
SMILES OC(=O)CCNC(=O)OCc1ccccc1
IUPAC Name 3-(phenylmethoxycarbonylamino)propanoic acid
Molecular Weight (g/mol) 223.23
Formula Weight 223.0845
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.