missing translation for 'onlineSavingsMsg'
Learn More

N-Carbethoxyphthalimide, 99+%

Product Code. 10095853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095853 250 g Glass bottle 250g
10727383 10 g Glass bottle 10g
10406910 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10095853 Supplier Thermo Scientific Acros Supplier No. 108312500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22509-74-6 | C11H9NO4 | 219.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22509-74-6
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.2
MDL Number MFCD00005893
InChI Key VRHAQNTWKSVEEC-UHFFFAOYSA-N
Synonym n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
PubChem CID 31187
IUPAC Name ethyl 1,3-dioxoisoindole-2-carboxylate
SMILES CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 82.0°C to 84.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 g
Beilstein 21, II, 357
Fieser 01,111; 05,91; 09,212
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Formula Weight 219.2
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Carbethoxyphthalimide
Show More Show Less
compliance-icons
Product Identifier
  • N-Carbethoxyphthalimide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.