missing translation for 'onlineSavingsMsg'
Learn More

N-Carbethoxy-4-piperidone, 98.5+%

Product Code. 10092200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092200 50 g Glass bottle 50g
10439090 10 g Glass bottle 10g
10628644 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092200 Supplier Thermo Scientific Acros Supplier No. 167240500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29976-53-2 | C8H13NO3 | 171.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29976-53-2
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.2
MDL Number MFCD00006188
InChI Key LUBGFMZTGFXIIN-UHFFFAOYSA-N
Synonym n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
PubChem CID 34767
IUPAC Name ethyl 4-oxopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1350g/mL
Boiling Point 95.0°C to 98.0°C (1.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4740 to 1.4760
Quantity 50 g
Specific Gravity 1.135
Solubility Information Solubility in water: slightly soluble
Formula Weight 171.2
Percent Purity 98.5+%
Physical Form Liquid
Chemical Name or Material N-Carbethoxy-4-piperidone
Show More Show Less
compliance-icons
Product Identifier
  • N-Carbethoxy-4-piperidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.