missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoylmaleimide, tech. 90%

Product Code. 11444654
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444654 50 g 50g
11434654 10 g 10g
11454654 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444654 Supplier Thermo Scientific Alfa Aesar Supplier No. A13685.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3345-50-4 | C5H4N2O3 | 140.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3345-50-4
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00014539
InChI Key BNPFHEFZJPVCCE-UHFFFAOYSA-N
Synonym n-carbamoylmaleimide, n-carbamylmaleimide, 2,5-dioxo-3-pyrroline-1-carboxamide, 2,5-dioxo-2,5-dihydro-1h-pyrrole-1-carboxamide, n-carbamoylmaleiimide, 3-pyrrolinecarboxamide, 2,5-dioxo, 3-pyrroline-2,5-dione, 1-carbamoyl, 1h-pyrrole-1-carboxamide, 2,5-dihydro-2,5-dioxo-9ci, 2,5-dihydro-2,5-dioxo-1h-pyrrole-1-carboxamide, 2,5-dioxoazolinecarboxamide
PubChem CID 76868
IUPAC Name 2,5-dioxopyrrole-1-carboxamide
SMILES C1=CC(=O)N(C1=O)C(=O)N

Specifications

Melting Point 157°C to 160°C
Quantity 50 g
Beilstein 117919
Formula Weight 140.1
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Carbamoylmaleimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.