missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoyl-DL-aspartic Acid, TRC

Product Code. 30352026
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30352026 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352026 Supplier Toronto Research Chemicals Supplier No. TRCC183680100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Carbamoyl-DL-aspartic Acid
CAS 923-37-5
Molecular Formula C5H8N2O5
Quantity 100 mg
Synonym N-(Aminocarbonyl)Aspartic Acid
InChI Formula InChI=1S/C5H8N2O5/c6-5(12)7-2(4(10)11)1-3(8)9/h2H,1H2,(H,8,9)(H,10,11)(H3,6,7,12)
SMILES NC(=O)NC(CC(=O)O)C(=O)O
IUPAC Name 2-(carbamoylamino)butanedioic acid
Molecular Weight (g/mol) 176.13
Formula Weight 176.0433
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.