missing translation for 'onlineSavingsMsg'
Learn More

n-Capric Acid, MP Biomedicals

Product Code. 11401836 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401836 100 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401836 Supplier MP Biomedicals Supplier No. 0520980450
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 334-48-5
Molecular Formula C10H20O2
Quantity 100 g
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILES CCCCCCCCCC(=O)O
IUPAC Name decanoic acid
Molecular Weight (g/mol) 172.268
PubChem CID 2969
ChEBI CHEBI:30813
Formula Weight 172.27
Chemical Name or Material n-Capric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.