missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-cadherin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22018-1-AP-20UL
This item is not returnable.
View return policy
Description
N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin′s function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.Specifications
| N-cadherin | |
| Polyclonal | |
| Unconjugated | |
| CDH2 | |
| Cadherin 2, CD325, CDH2, CDHN, CDw325, N cadherin, NCAD, N-cadherin, Neural cadherin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1000, 12558, 83501 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15116, P19022, Q9Z1Y3 | |
| Cdh2 | |
| N-cadherin Fusion Protein Ag16792 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction