Learn More
N-cadherin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22018-1-AP-150UL
 View more versions of this product
Product Code. 16871555
  • £349.00 / 150µL
Description
N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin′s function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.
Specifications
Specifications
N-cadherin
Polyclonal
Unconjugated
CDH2
Cadherin 2, CD325, CDH2, CDHN, CDw325, N cadherin, NCAD, N-cadherin, Neural cadherin
Rabbit
Antigen Affinity Chromatography
RUO
1000, 12558, 83501
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15116, P19022, Q9Z1Y3
Cdh2
N-cadherin Fusion Protein Ag16792
150 μL
Primary
Human, Mouse, Rat