missing translation for 'onlineSavingsMsg'
Learn More

N-Cadherin Antibody (8C11), DyLight 405, Novus Biologicals™

Product Code. 18411093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411093 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411093 Supplier Novus Biologicals Supplier No. NBP254523V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-Cadherin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8C11
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin
Host Species Mouse
Immunogen Recombinant human N-Cadherin extracellular domain (Uniprot: P19022 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.