missing translation for 'onlineSavingsMsg'
Learn More

N-Cadherin Antibody (13A9), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18403205 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403205 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403205 Supplier Novus Biologicals Supplier No. NBP148309MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone 13A9
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin
Host Species Mouse
Immunogen This N-Cadherin Antibody (13A9) was developed against the cytoplasmic domain of human N Cadherin [Swiss-Prot# P19022].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.