missing translation for 'onlineSavingsMsg'
Learn More

N-butyryl-L-Homoserine lactone, TRC

Product Code. 30393278
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393278 10 mg 10mg
30389270 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393278 Supplier Toronto Research Chemicals Supplier No. TRCH61512810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67605-85-0
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.19
Synonym N-[(3S)-Tetrahydro-2-oxo-3-furanyl]butanamide, N-Butanoyl-L-homoserine lactone, N-Butyryl-L-homoserine lactone, PAI, (S)-N-(Tetrahydro-2-oxo-3-furanyl)-butanamide
IUPAC Name N-[(3S)-2-oxooxolan-3-yl]butanamide
SMILES CCCC(=O)N[C@H]1CCOC1=O

Specifications

InChI Formula InChI=1S/C8H13NO3/c1-2-3-7(10)9-6-4-5-12-8(6)11/h6H,2-5H2,1H3,(H,9,10)/t6-/m0/s1
Quantity 10 mg
Formula Weight 171.0895
Shipping Condition Room Temperature
Chemical Name or Material N-butyryl-L-Homoserine lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.