missing translation for 'onlineSavingsMsg'
Learn More

n-Butyronitrile, 99%

Product Code. 10305840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10305840 2.5 L 2.5 L
10577743 100 mL 100mL
10211463 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10305840 Supplier Thermo Scientific Supplier No. 108140025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-74-0 | C4H7N | 69.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-74-0
MDL Number MFCD00001968
InChI Key KVNRLNFWIYMESJ-UHFFFAOYSA-N
Synonym butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile
PubChem CID 8008
ChEBI CHEBI:51937
IUPAC Name butanenitrile
SMILES CCCC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -112.0°C
Density 0.7900g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7N
Linear Formula CH3CH2CH2CN
Refractive Index 1.3830 to 1.3850
Quantity 2.5 L
Beilstein 02, II, 252
Merck Index 15, 1601
Specific Gravity 0.79
Solubility Information Solubility in water: 33g/L (25°C). Other solubilities: miscible with alcohol,ether and dimethylformamide
Molecular Weight (g/mol) 69.11
Viscosity 0.57 mPa.s (20°C)
Formula Weight 69.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Butyronitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Butyronitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: n-Butanenitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.