missing translation for 'onlineSavingsMsg'
Learn More

N-Butyric Acid 0.995 MP Biomedicals

Product Code. 11338789 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338789 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11338789 Supplier MP Biomedicals Supplier No. 0210116980
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 107-92-6
Density 3.0 (air = 1)
Quantity 100 g
Assay Percent Range 0.995
Molecular Formula C4H8O2
MDL Number MFCD00002814
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
SMILES CCCC(O)=O
IUPAC Name butanoic acid
Molecular Weight (g/mol) 88.11
PubChem CID 264
ChEBI CHEBI:30772
Formula Weight 88.11
Chemical Name or Material N-Butyric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.