missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltriphenylphosphonium Bromide, TRC

Product Code. 30339233
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
missing translation for 'unitSize'
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339233 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30339233 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB69380510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Butyltriphenylphosphonium Bromide
CAS 1779-51-7
Molecular Formula C22H24BrP
Quantity 10 g
Synonym Butyltriphenylphosphonium Bromide, B 0970, BTPPBr, Butyl Bromide triphenylphosphine Salt, Hishicolin BTPPBr, NSC 59684, Triphenylbutylphosphonium Bromide, n-Butyltriphenylphosphonium Bromide
InChI Formula InChI=1S/C22H24P.BrH/c1-2-3-19-23(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22;/h4-18H,2-3,19H2,1H3;1H/q+1;/p-1
SMILES [Br-].CCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name butyl(triphenyl)phosphanium;bromide
Molecular Weight (g/mol) 399.3
Formula Weight 398.0799
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.