missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltrichlorosilane, 97+%

Product Code. 11484310
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484310 100 g 100g
11494310 5 g 5g
11474310 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484310 Supplier Thermo Scientific Alfa Aesar Supplier No. A11256.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7521-80-4 | C4H9Cl3Si | 191.551 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7521-80-4
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.551
MDL Number MFCD00013605
InChI Key FQEKAFQSVPLXON-UHFFFAOYSA-N
Synonym silane, butyltrichloro, n-butyltrichlorosilane, trichlorobutylsilane, unii-zq9o7lyo00, zq9o7lyo00, butylsilicon trichloride, n-butyl trichlorosilane, butyl trichlorosilane, hsdb 283, butyl-trichlorosilane
PubChem CID 24142
IUPAC Name butyl(trichloro)silane
SMILES CCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 142°C to 143°C
Quantity 100 g
Linear Formula CH3(CH2)3SiCl3
UN Number UN1747
Beilstein 1736032
Flash Point 45°C (113°F)
Formula Weight 191.56
Percent Purity ≥97%
Odor Acrid
Refractive Index 1.437
Sensitivity Moisture sensitive
Density 1.16
Chemical Name or Material n-Butyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.