missing translation for 'onlineSavingsMsg'
Learn More

n-Butylidenephthalide, (E)+(Z), 95%

Product Code. p-7080707
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11494143

Brand: Thermo Scientific Alfa Aesar A10353.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 551-08-6 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-08-6
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00047319,MFCD00047319
InChI Key WMBOCUXXNSOQHM-DHZHZOJOSA-N
Synonym ligusticum lactone, butylidene phthalide, n-butylidene phthalide, 3-butylidene-1 3h-isobenzofuranone, 3-butylidenephthalide, phthalide, 3-butylidene, fema no. 3333, bdph, 3-butylidene phthalide, 1 3h-isobenzofuranone, 3-butylidene
PubChem CID 5352899
IUPAC Name (3E)-3-butylidene-2-benzofuran-1-one
SMILES CCC\C=C1\OC(=O)C2=CC=CC=C12

Specifications

Density 1.103
Boiling Point 108°C to 109°C (0.1 mmHg)
Flash Point 132°C (269°F)
Refractive Index 1.577
Quantity 10 g
Beilstein 5145
Formula Weight 188.23
Percent Purity 95%
Chemical Name or Material n-Butylidenephthalide, (E)+(Z)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.