missing translation for 'onlineSavingsMsg'
Learn More

n-Butylcyclohexane, 99%

Product Code. 11392736
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11392736

Brand: Thermo Scientific Alfa Aesar L08845.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1678-93-9 | C10H20 | 140.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1678-93-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00001530
InChI Key GGBJHURWWWLEQH-UHFFFAOYSA-N
Synonym n-butylcyclohexane, 1-cyclohexylbutane, cyclohexane, butyl, butyl-cyclohexane, butane, 1-cyclohexyl, n-butyl-cyclohexane, 1-cyclohexyl-butane, cyclohexane, n-butyl, acmc-20aj43, butylcyclohexane
PubChem CID 15506
IUPAC Name butylcyclohexane
SMILES CCCCC1CCCCC1

Specifications

Melting Point -78°C
Density 0.818
Boiling Point 178°C to 180°C
Flash Point 50°C (122°F)
Refractive Index 1.441
Quantity 50 g
UN Number UN3295
Beilstein 1900631
Formula Weight 140.27
Percent Purity 99%
Chemical Name or Material n-Butylcyclohexane
Show More Show Less
compliance-icons
Product Identifier
  • n-Butylcyclohexane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Butylcyclohexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.