missing translation for 'onlineSavingsMsg'
Learn More

n-Butylboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10056043
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
4426-47-5
C4H11BO2
101.94
MFCD00002106
QPKFVRWIISEVCW-UHFFFAOYSA-N
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane
20479
butylboronic acid
CCCCB(O)O
4426-47-5
C4H11BO2
101.94
MFCD00002106
QPKFVRWIISEVCW-UHFFFAOYSA-N
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane
20479
butylboronic acid
CCCCB(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 94.0°C to 96.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)3B(OH)2
Quantity 25 g
Solubility Information Solubility in water:+/- 2.5% in water
Formula Weight 101.94
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material n-Butylboronic acid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • n-Butylboronic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'