missing translation for 'onlineSavingsMsg'
Learn More

n-Butylbenzene, 99%, Thermo Scientific Chemicals

Catalog Number 11404293
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
104-51-8
C10H14
134.22
MFCD00009463
OCKPCBLVNKHBMX-UHFFFAOYSA-N
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
7705
CHEBI:44194
butylbenzene
CCCCC1=CC=CC=C1
This item is not returnable. View return policy
104-51-8
C10H14
134.22
MFCD00009463
OCKPCBLVNKHBMX-UHFFFAOYSA-N
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
7705
CHEBI:44194
butylbenzene
CCCCC1=CC=CC=C1

n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

Solubility
Sparingly soluble in water (0.012g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -88°C
Density 0.86
Boiling Point 181°C to 183°C
Flash Point 59°C (138°F)
Odor Aromatic
Refractive Index 1.489
Quantity 50 g
UN Number UN2709
Beilstein 1903395
Merck Index 14,1549
Solubility Information Sparingly soluble in water (0.012g/L).
Formula Weight 134.22
Percent Purity 99%
Chemical Name or Material n-Butylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'