missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl propionate, 99%

Product Code. p-7024878
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11460651

Brand: Thermo Scientific Alfa Aesar B20125.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-01-2 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-01-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009448
InChI Key BTMVHUNTONAYDX-UHFFFAOYSA-N
Synonym butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
PubChem CID 11529
IUPAC Name butyl propanoate
SMILES CCCCOC(=O)CC

Specifications

Density 0.87
Boiling Point 144°C to 146°C
Flash Point 38°C (101°F)
Refractive Index 1.402
Quantity 100 g
UN Number UN3272
Merck Index 14,1587
Formula Weight 130.19
Percent Purity 99%
Chemical Name or Material n-Butyl propionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.