missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11489366
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11489366

Brand: Thermo Scientific Alfa Aesar A19068.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.

Solubility
Miscible with ethyl ether and ethanol. Immiscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-88-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00009444
InChI Key SOGAXMICEFXMKE-UHFFFAOYSA-N
Synonym butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
PubChem CID 7354
IUPAC Name butyl 2-methylprop-2-enoate
SMILES CCCCOC(=O)C(C)=C

Specifications

Melting Point -75°C
Density 0.894
Boiling Point 160°C to 163°C
Flash Point 49°C (120°F)
Odor Fruit-like
Refractive Index 1.424
Quantity 2500 mL
UN Number UN2227
Beilstein 773960
Solubility Information Miscible with ethyl ether and ethanol. Immiscible with water.
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material n-Butyl methacrylate, stabilized with 15 to 20 ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.