missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11479366
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479366 500 mL 500mL
11489366 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479366 Supplier Thermo Scientific Alfa Aesar Supplier No. A19068.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.

Solubility
Miscible with ethyl ether and ethanol. Immiscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-88-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00009444
InChI Key SOGAXMICEFXMKE-UHFFFAOYSA-N
Synonym butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
PubChem CID 7354
IUPAC Name butyl 2-methylprop-2-enoate
SMILES CCCCOC(=O)C(C)=C

Specifications

Melting Point -75°C
Density 0.894
Boiling Point 160°C to 163°C
Flash Point 49°C (120°F)
Odor Fruit-like
Refractive Index 1.424
Quantity 500 mL
UN Number UN2227
Beilstein 773960
Solubility Information Miscible with ethyl ether and ethanol. Immiscible with water.
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material n-Butyl methacrylate, stabilized with 15 to 20 ppm 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Butyl methacrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Butyl methacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.