missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl lactate, 99%

Product Code. 10300944 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10300944 2.5 L Glass bottle 2.5 L
10154530 25 mL Glass bottle 25mL
10743051 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10300944 Supplier Thermo Scientific Acros Supplier No. 250180025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138-22-7 | C7H14O3 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-22-7
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.19
MDL Number MFCD00004519
InChI Key MRABAEUHTLLEML-UHFFFAOYNA-N
Synonym butyl lactate, n-butyl lactate, lactic acid, butyl ester, propanoic acid, 2-hydroxy-, butyl ester, butyllactate, 2-hydroxypropanoic acid butyl ester, butylester kyseliny mlecne, butyl alpha-hydroxypropionate, butyl lactate natural, fema no. 2205
PubChem CID 8738
IUPAC Name butyl 2-hydroxypropanoate
SMILES CCCCOC(=O)C(C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless
Density 0.9800g/mL
Boiling Point 185.0°C to 187.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4200 to 1.4220
Quantity 2.5 L
Specific Gravity 0.98
Solubility Information Solubility in water: 42g/L (25 c). Other solubilities: hydrolyses in acids and alkalies,miscible with ether alcohol
Formula Weight 146.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Butyl lactate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.