Learn More
n-Butyl L-lactate, 97+%
Description
Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Fenofibrate was formulated with n-butyl L-lactate, Tween 80, and a number of cosurfactants, diluted with the water phase (either water or saccharidesolution) and then subjected to a freeze-drying (FD) process to obtain SMEPMS nanosized.
Solubility
Soluble in alcohol and water(42 g/L (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Density | 0.984 |
| Boiling Point | 185°C to 187°C |
| Flash Point | 69°C (157°F) |
| Refractive Index | 1.421 |
| Quantity | 5000 g |
| Synonym | butyl 2s-2-hydroxypropanoate, unii-33glz553p8, butyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, butyl ester, 2s, butyl l-lactate, --butyl l-lactate, propanoic acid,2-hydroxy-, butyl ester, 2s, +-butyl lactate, butyl-l-lactate, butyl lactate, s |
| Solubility Information | Soluble in alcohol and water(42g/L (25°C). |
| Formula Weight | 146.19 |
| Percent Purity | ≥97% |
| Chemical Name or Material | n-Butyl L-lactate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.