missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl heptafluorobutyrate, 97%, Thermo Scientific™

Product Code. 11460517
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460517 10 g 10g
11470517 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460517 Supplier Thermo Scientific Alfa Aesar Supplier No. B22400.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1559-07-5
Molecular Formula C8H9F7O2
Molecular Weight (g/mol) 270.147
MDL Number MFCD00039240
InChI Key YDXXJZKOOOJVEE-UHFFFAOYSA-N
Synonym n-butyl heptafluorobutyrate, butyric acid, heptafluoro-, butyl ester, butyl heptafluorobutanoate, heptafluorobutyric acid, n-butyl ester, n-butylheptafluorobutyrate, butyl perfluorobutanoat, butyl perfluorobutyrate, butylheptafluorobutyrate, butyl heptafluorobutyrate, n-butyl perfluoro-n-butyrate
PubChem CID 137079
IUPAC Name butyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.296
Boiling Point 131°C to 133°C
Linear Formula CF3CF2CF2CO2(CH2)3CH3
Refractive Index 1.327
Quantity 10 g
Beilstein 1798327
Formula Weight 270.15
Percent Purity 97%
Chemical Name or Material n-Butyl heptafluorobutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.