missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl formate, 97%

Product Code. p-7021520
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
This item is not returnable. View return policy

Product Code. 11438760

Brand: Thermo Scientific Alfa Aesar A13882.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-84-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 592-84-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00003296
InChI Key NMJJFJNHVMGPGM-UHFFFAOYSA-N
Synonym n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
PubChem CID 11614
IUPAC Name butyl formate
SMILES CCCCOC=O

Tekniske data

Melting Point -92°C
Boiling Point 105°C to 107°C
Flash Point 18°C (64°F)
Quantity 50 mL
UN Number UN1128
Beilstein 1742108
Formula Weight 102.13
Percent Purity 97%
Chemical Name or Material n-Butyl formate

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.