missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl ethyl ether, 98+%, Thermo Scientific™

Product Code. 11469096
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11469096

Brand: Thermo Scientific Alfa Aesar A17963.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-81-9
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00009455
InChI Key PZHIWRCQKBBTOW-UHFFFAOYSA-N
Synonym butyl ethyl ether, ethyl butyl ether, butane, 1-ethoxy, ether, butyl ethyl, n-butyl ethyl ether, ether ethylbutylique, 1-butyl ethyl ether, unii-no396r19pj, ether ethylbutylique french, ethyl n-butyl ether
PubChem CID 12355
IUPAC Name 1-ethoxybutane
SMILES CCCCOCC

Specifications

Melting Point -124°C
Density 0.75
Boiling Point 91°C to 92°C
Flash Point −6°C (21°F)
Refractive Index 1.382
Quantity 10 g
UN Number UN1179
Beilstein 1731323
Solubility Information Slightly soluble in water
Formula Weight 102.18
Percent Purity ≥98%
Chemical Name or Material n-Butyl ethyl ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.