missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl chloroformate, 98%

Product Code. 11448760
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448760 100 g 100g
11445846 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448760 Supplier Thermo Scientific Alfa Aesar Supplier No. B22215.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-34-7 | C5H9ClO2 | 136.575 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-34-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00000650
InChI Key NRDQFWXVTPZZAZ-UHFFFAOYSA-N
Synonym butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate
PubChem CID 61140
IUPAC Name butyl carbonochloridate
SMILES CCCCOC(=O)Cl

Specifications

Melting Point <-70°C
Density 1.06
Boiling Point 141°C to 143°C
Flash Point 39°C (102°F)
Odor Acrid
Refractive Index 1.413
Quantity 100 g
UN Number UN2743
Beilstein 605635
Sensitivity Moisture sensitive
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material n-Butyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Butyl chloroformate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.